Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl sulfite, 98%, Thermo Scientific™

Catalog No. AC209930100
Change view
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC209930100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC209930100 Supplier Thermo Scientific Chemicals Supplier No. 209930100

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 623-81-4
Molecular Formula C4H10O3S
Molecular Weight (g/mol) 138.18
MDL Number MFCD00009098
InChI Key NVJBFARDFTXOTO-UHFFFAOYSA-N
Synonym sulfurous acid, diethyl ester, ethyl sulfite, diethyl sulphite, ethyl sulfite, et2so3, diethylsulfite, ethoxysulfinyl ethoxy, diethylsulfit, sulfurous acid diethyl, diethyl sulfite, sulfurous acid diethyl ester
PubChem CID 12197
IUPAC Name diethyl sulfite
SMILES CCOS(=O)OCC

Specifications

Color Colorless
Density 1.0800g/mL
Boiling Point 158°C to 160°C
Flash Point 53°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4130 to 1.415
Linear Formula (C2H5O)2SO
Quantity 10 g
Beilstein 01, 325
Specific Gravity 1.08
Formula Weight 138.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Diethyl sulfite, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 210-815-5

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.