accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOBr

CAS RN 6232-88-8

IUPAC Name: 4-(bromomethyl)benzoic acid
Synonyms: 4-(bromomethyl)benzoic acid
Molecular Weight (g/mol): 215.05
Molecular Formula: C8H7BrO2
InChi Key: CQQSQBRPAJSTFB-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(CBr)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (5)
  • (3)
Percent Purity 
  • (2)
  • (9)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (2)
  • (6)
  • (3)
  • (5)
  • (3)
  • (2)
  • (9)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (5)