CAS RN 62325-30-8
CAS RN 62325-30-8
IUPAC Name:
2,7-dibromophenanthrene
Synonyms:
2,7-dibromophenanthrene
Molecular Weight (g/mol):
336.03
Molecular Formula:
C14H8Br2
InChi Key:
NGDWMVTZPZDKPM-UHFFFAOYSA-N
SMILES:
BrC1=CC=C2C(C=CC3=CC(Br)=CC=C23)=C1
1
–
1
of
1
results
1
2,7-Dibromophenanthrene 98.0+%, TCI America™
CAS: 62325-30-8 Molecular Formula: C14H8Br2 Molecular Weight (g/mol): 336.026 MDL Number: MFCD09909861 InChI Key: NGDWMVTZPZDKPM-UHFFFAOYSA-N PubChem CID: 11175073 IUPAC Name: 2,7-dibromophenanthrene SMILES: C1=CC2=C(C=CC3=C2C=CC(=C3)Br)C=C1Br