Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,7-Dibromophenanthrene 98.0+%, TCI America™
SDP

Catalog No. D5050200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D5050200MG 200 mg
D50501G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D5050200MG Supplier TCI America Supplier No. D5050200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 62325-30-8
Molecular Formula C14H8Br2
Molecular Weight (g/mol) 336.026
MDL Number MFCD09909861
InChI Key NGDWMVTZPZDKPM-UHFFFAOYSA-N
PubChem CID 11175073
IUPAC Name 2,7-dibromophenanthrene
SMILES C1=CC2=C(C=CC3=C2C=CC(=C3)Br)C=C1Br

Specifications

Melting Point 205°C
Color White-Yellow
Quantity 200 mg
Formula Weight 336.03
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,7-Dibromophenanthrene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.