accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 62336-24-7

IUPAC Name: (2-bromophenyl)diphenylphosphane
Synonyms: (2-bromophenyl)diphenylphosphane
Molecular Weight (g/mol): 341.19
Molecular Formula: C18H14BrP
InChi Key: XIONUQPOXCUMMB-UHFFFAOYSA-N
SMILES: BrC1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results