accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 62365-34-8

IUPAC Name: 1-(ethyldimethylsilyl)-1H-imidazole
Synonyms: 1-(ethyldimethylsilyl)imidazole
Molecular Weight (g/mol): 154.29
Molecular Formula: C7H14N2Si
InChi Key: HMMXVNBTJZQGTI-UHFFFAOYSA-N
SMILES: CC[Si](C)(C)N1C=CN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results