Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(Dimethylethylsilyl)imidazole 98.0+%, TCI America™
SDP

Catalog No. D15165G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D15165G 5 g
D15161G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D15165G Supplier TCI America Supplier No. D15165G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 62365-34-8
Molecular Formula C7H14N2Si
Molecular Weight (g/mol) 154.29
MDL Number MFCD00059723
InChI Key HMMXVNBTJZQGTI-UHFFFAOYSA-N
PubChem CID 173854
IUPAC Name 1-(ethyldimethylsilyl)-1H-imidazole
SMILES CC[Si](C)(C)N1C=CN=C1

Specifications

Color Colorless
Boiling Point 80°C
Quantity 5 g
Formula Weight 154.29
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 1-(Dimethylethylsilyl)imidazole [Dimethylethylsilylating Agent]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.