accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6238-12-6

IUPAC Name: 7-methoxy-4-methylquinoline
Synonyms: 7-methoxy-4-methylquinoline
Molecular Weight (g/mol): 173.22
Molecular Formula: C11H11NO
InChi Key: LVJHDVPCJRWNNZ-UHFFFAOYSA-N
SMILES: COC1=CC=C2C(C)=CC=NC2=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results