Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(+)-Citramalic Acid 98+%, Thermo Scientific™

Catalog No. AC276660050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC276660050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC276660050 Supplier Thermo Scientific Chemicals Supplier No. 276660050

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6238-12-6
Molecular Formula C11H11NO
Molecular Weight (g/mol) 173.22
InChI Key LVJHDVPCJRWNNZ-UHFFFAOYSA-N
Synonym 4-methyl-7-methoxyquinoline, quinoline, 7-methoxy-4-methyl, 4-nitro-phenyl-p-tolyl-methanone
PubChem CID 13685551
IUPAC Name 7-methoxy-4-methylquinoline
SMILES CC1=C2C=CC(=CC2=NC=C1)OC

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 106°C to 110°C
Color White
Infrared Spectrum Authentic
Linear Formula HO2CCH2C(CH3)(OH)CO2H
Quantity 5 g
Beilstein 03, 443
Merck Index 14, 2323
Specific Rotation +21° to +25° (20°C, 589nm) (c=2, H2O)
Formula Weight 148.12
Percent Purity ≥98.0%
Physical Form Fine Crystalline Powder
Chemical Name or Material (S)-(+)-Citramalic acid, 98+%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.