accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHCH3

CAS RN 624-08-8

IUPAC Name: heptadecan-9-ol
Synonyms: 9-heptadecanol
Molecular Weight (g/mol): 256.47
Molecular Formula: C17H36O
InChi Key: WTWWTKPAEZQYPW-UHFFFAOYSA-N
SMILES: CCCCCCCCC(O)CCCCCCCC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results