Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Heptadecanol 95.0+%, TCI America™
SDP

Catalog No. H14011G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H14011G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H14011G Supplier TCI America Supplier No. H14011G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 624-08-8
Molecular Formula C17H36O
Molecular Weight (g/mol) 256.474
MDL Number MFCD00046734
InChI Key WTWWTKPAEZQYPW-UHFFFAOYSA-N
PubChem CID 136435
IUPAC Name heptadecan-9-ol
SMILES CCCCCCCCC(CCCCCCCC)O

Specifications

Melting Point 60°C
Color White
Quantity 1 g
Formula Weight 256.47
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9-Heptadecanol
EINECSNumber (2)-0217
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.