CAS RN 624-13-5
CAS RN 624-13-5
IUPAC Name:
propyl octanoate
Synonyms:
propyl octanoate
Molecular Weight (g/mol):
186.3
Molecular Formula:
C11H22O2
InChi Key:
IDHBLVYDNJDWNO-UHFFFAOYSA-N
SMILES:
CCCCCCCC(=O)OCCC
1
–
1
of
1
results
1
Propyl n-Octanoate 98.0+%, TCI America™

CAS: 624-13-5 Molecular Formula: C11H22O2 Molecular Weight (g/mol): 186.30 MDL Number: MFCD00056317 InChI Key: IDHBLVYDNJDWNO-UHFFFAOYSA-N Synonym: n-Octanoic Acid Propyl Ester PubChem CID: 69351 ChEBI: CHEBI:87374 IUPAC Name: propyl octanoate SMILES: CCCCCCCC(=O)OCCC