Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propyl n-Octanoate 98.0+%, TCI America™
SDP

Catalog No. P211325ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P211325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P211325ML Supplier TCI America Supplier No. P211325ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 624-13-5
Molecular Formula C11H22O2
Molecular Weight (g/mol) 186.30
MDL Number MFCD00056317
InChI Key IDHBLVYDNJDWNO-UHFFFAOYSA-N
Synonym n-Octanoic Acid Propyl Ester
PubChem CID 69351
ChEBI CHEBI:87374
IUPAC Name propyl octanoate
SMILES CCCCCCCC(=O)OCCC

Specifications

Color Yellow
Boiling Point 226°C
Quantity 25 mL
Formula Weight 186.30
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Propyl n-Octanoate
EINECSNumber (2)-0782
RTECSNumber RH0786500
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.