accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 624-75-9

IUPAC Name: 2-iodoacetonitrile
Synonyms: iodoacetonitrile
Molecular Weight (g/mol): 166.95
Molecular Formula: C2H2IN
InChi Key: VODKOOOHHCAWFR-UHFFFAOYSA-N
SMILES: ICC#N
Boiling Point 
  • (2)
  • (2)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)