accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 624-78-2

IUPAC Name: ethyl(methyl)amine
Synonyms: methylethylamine
Molecular Weight (g/mol): 59.11
Molecular Formula: C3H9N
InChi Key: LIWAQLJGPBVORC-UHFFFAOYSA-N
SMILES: CCNC
Boiling Point 
  • (2)
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (4)
Quantity 
  • (4)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (1)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (4)
  • (4)
  • (4)