Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N-Ethylmethylamine

Catalog No. 502891511
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-289-1511 25 g
29-114-55G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-289-1511 Supplier Sigma Aldrich Supplier No. 29114525G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

The kinetics of reaction of guanosine 5α-phospho-2-methylimidazolide with N-ethylmethylamine has been studied in water at 37C.N-Ethylmethylamine has been used in the synthesis of N-ethylmethylacrylamide via reaction with acryloyl chloride.
TRUSTED_SUSTAINABILITY

Specifications

CAS 624-78-2
Density 0.688 g/mL (at 25°C (literature))
Boiling Point 36°C to 37°C (lit.)
Molecular Formula C3H9N
Refractive Index n20/D 1.374 (literature)
Linear Formula C2H5NHCH3
MDL Number MFCD00009030
Quantity 25 g
Synonym N-Methylethylamine; NMEA
Molecular Weight (g/mol) 59.11
Percent Purity 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.