accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOONHOOOHO

CAS RN 6245-75-6

IUPAC Name: 3-[bis(carboxymethyl)amino]propanoic acid
Synonyms: β-alanine diacetic acid
Molecular Weight (g/mol): 205.17
Molecular Formula: C7H11NO6
InChi Key: UWRBFYBQPCJRRL-UHFFFAOYSA-N
SMILES: OC(=O)CCN(CC(O)=O)CC(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results