Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(2-Carboxyethyl)iminodiacetic Acid 98.0+%, TCI America™
SDP

Catalog No. N04085G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N04085G 5 g
N040825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N04085G Supplier TCI America Supplier No. N04085G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6245-75-6
Molecular Formula C7H11NO6
Molecular Weight (g/mol) 205.17
MDL Number MFCD00059634
InChI Key UWRBFYBQPCJRRL-UHFFFAOYSA-N
Synonym n-2-carboxyethyl iminodiacetic acid, beta-alanine diacetic acid, unii-kou355f9z2, n,n-bis carboxymethyl-, a-alanine, .beta.-alanine, n,n-bis carboxymethyl, 3-bis carboxymethyl amino propanoic acid, n-2-carboxyethyl dimethylsulfonium chloride, n,n-bis carboxymethyl-beta-alanine, beta-alaninediacetic acid, nitrilodiacetic-propionic acid
PubChem CID 535795
IUPAC Name 3-[bis(carboxymethyl)amino]propanoic acid
SMILES OC(=O)CCN(CC(O)=O)CC(O)=O

Specifications

Melting Point 195°C
Color White
Quantity 5 g
Formula Weight 205.17
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-(2-Carboxyethyl)iminodiacetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.