accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 62484-16-6

IUPAC Name: 6-methyl-1,2,3,4-tetrahydroquinazoline-2,4-dione
Synonyms: quinazoline-2,4-dione,6-ME
Molecular Weight (g/mol): 176.18
Molecular Formula: C9H8N2O2
InChi Key: OXVPNGBFUAIXTE-UHFFFAOYSA-N
SMILES: CC1=CC=C2NC(=O)NC(=O)C2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results