accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 625-45-6

IUPAC Name: 2-methoxyacetic acid
Synonyms: methoxyacetic acid
Molecular Weight (g/mol): 90.08
Molecular Formula: C3H6O3
InChi Key: RMIODHQZRUFFFF-UHFFFAOYSA-N
SMILES: COCC(O)=O
Molecular Weight (g/mol) 
  • (6)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (6)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (3)
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (2)
  • (6)
  • (6)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (6)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)
  • (1)
  • (5)