accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 625-50-3

IUPAC Name: N-ethylacetamide
Synonyms: acetamide, N-ethyl-
Molecular Weight (g/mol): 87.12
Molecular Formula: C4H9NO
InChi Key: PMDCZENCAXMSOU-UHFFFAOYSA-N
SMILES: CCNC(C)=O
Boiling Point 
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)