Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Ethylacetamide, 99%, Thermo Scientific™

Catalog No. AAL1030018
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1030018 50 g
AAL1030009 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1030018 Supplier Thermo Scientific Chemicals Supplier No. L1030018

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
N-ethylacetamide is a suitable nylon model for mechanistic studies of dye fading. N-Ethylacetamide was used to investigate propylene carbonate and a mixture of N-methylformamide and N-ethylacetamide by broadband dielectric and mechanical shear spectroscopy. It was used in determination of reaction products of OH-oxidation of N-methylpyrrolidone under atmospheric conditions.

Solubility
Fully misicible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 625-50-3
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.12
MDL Number MFCD00009029
InChI Key PMDCZENCAXMSOU-UHFFFAOYSA-N
Synonym acetamide, n-ethyl, acetamidoethane, acetoethylamide, ethylacetamide, n-aethylacetamid, n-acetylethylamine, n-aethylacetamid german, n-ethyl-acetamide, unii-m9y95ze6j9, 1-acetamidoethyl
PubChem CID 12253
ChEBI CHEBI:87370
IUPAC Name N-ethylacetamide
SMILES CCNC(C)=O

Specifications

Melting Point -32°C
Density 0.924
Boiling Point 206°C to 208°C
Flash Point 106°C (222°F)
Refractive Index 1.433
Quantity 50 g
Beilstein 969292
Solubility Information Fully misicible in water.
Formula Weight 87.12
Percent Purity 99%
Chemical Name or Material N-Ethylacetamide
Show More Show Less
EINECSNumber 210-896-7
RTECSNumber AB8500000
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.