accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONHCH3O

CAS RN 625-77-4

IUPAC Name: N-acetylacetamide
Synonyms: diacetamide
Molecular Weight (g/mol): 101.11
Molecular Formula: C4H7NO2
InChi Key: ZSBDPRIWBYHIAF-UHFFFAOYSA-N
SMILES: CC(=O)NC(C)=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Boiling Point 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results