Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diacetamide 98%, Thermo Scientific™

Catalog No. AC112000250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC112000250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC112000250 Supplier Thermo Scientific Chemicals Supplier No. 112000250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 625-77-4
Molecular Formula C4H7NO2
Molecular Weight (g/mol) 101.105
MDL Number MFCD00008679
InChI Key ZSBDPRIWBYHIAF-UHFFFAOYSA-N
Synonym diacetamide, acetamide, n-acetyl, diacetylamine, n-acetyl-acetamide, ch3c o nhc o ch3, bisacetylamine, nn-diacetylamine, n-acetylacetamide, acetamide,n-acetyl
PubChem CID 12263
IUPAC Name N-acetylacetamide
SMILES CC(=O)NC(=O)C

Specifications

Melting Point 74°C to 78°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Quantity 25 g
Beilstein 02, 181
Formula Weight 101.1
Percent Purity ≥97.5%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material Diacetamide, 98%
EINECSNumber 210-910-1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.