accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHOOH

CAS RN 6250-70-0

IUPAC Name: nonadecanedioic acid
Synonyms: nonadecanedioic acid
Molecular Weight (g/mol): 328.49
Molecular Formula: C19H36O4
InChi Key: IFAWYXIHOVRGHQ-UHFFFAOYSA-N
SMILES: OC(=O)CCCCCCCCCCCCCCCCCC(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results