Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nonadecanedioic Acid 98.0+%, TCI America™
SDP

Catalog No. N06635G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N06635G 5 g
N06631G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N06635G Supplier TCI America Supplier No. N06635G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6250-70-0
Molecular Formula C19H36O4
Molecular Weight (g/mol) 328.49
MDL Number MFCD00039536
InChI Key IFAWYXIHOVRGHQ-UHFFFAOYSA-N
Synonym 1,17-Heptadecanedicarboxylic Acid
PubChem CID 12572015
IUPAC Name nonadecanedioic acid
SMILES OC(=O)CCCCCCCCCCCCCCCCCC(O)=O

Specifications

Quantity 5 g
Formula Weight 328.49
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Nonadecanedioic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.