accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNNN

CAS RN 6251-01-0

IUPAC Name: 2-[6-(dicyanomethylidene)-2,6-dihydronaphthalen-2-ylidene]propanedinitrile
Synonyms: 2-[6-(dicyanomethylidene)naphthalen-2-ylidene]propanedinitrile
Molecular Weight (g/mol): 254.25
Molecular Formula: C16H6N4
InChi Key: JLTPSDHKZGWXTD-UHFFFAOYSA-N
SMILES: N#CC(C#N)=C1C=CC2=CC(C=CC2=C1)=C(C#N)C#N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results