Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

11,11,12,12-Tetracyanonaphtho-2,6-quinodimethane 98.0+%, TCI America™
SDP

Catalog No. T1246100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1246100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T1246100MG Supplier TCI America Supplier No. T1246100MG
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6251-01-0
Molecular Formula C16H6N4
Molecular Weight (g/mol) 254.252
MDL Number MFCD00059115
InChI Key JLTPSDHKZGWXTD-UHFFFAOYSA-N
Synonym TNAP
PubChem CID 138704
IUPAC Name 2-[6-(dicyanomethylidene)naphthalen-2-ylidene]propanedinitrile
SMILES C1=CC(=C(C#N)C#N)C=C2C1=CC(=C(C#N)C#N)C=C2

Specifications

Color Blue-Purple
Quantity 100 mg
UN Number 3439
Formula Weight 254.25
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 11,11,12,12-Tetracyanonaphtho-2,6-quinodimethane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.