accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 62538-21-0

IUPAC Name: 2-(4-chlorophenyl)-3-oxopropanenitrile
Synonyms: 2-(4-chlorophenyl)-3-oxopropanenitrile
Molecular Weight (g/mol): 179.6
Molecular Formula: C9H6ClNO
InChi Key: DAEXXSXAEMFPHQ-UHFFFAOYNA-N
SMILES: ClC1=CC=C(C=C1)C(C=O)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results