Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(4-Chlorophenyl)-3-oxopropanenitrile, 95%, Thermo Scientific™

Catalog No. SPB01054DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SPB01054DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SPB01054DA Supplier Thermo Scientific Chemicals Supplier No. SPB01054DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 62538-21-0
Molecular Formula C9H6ClNO
Molecular Weight (g/mol) 179.60
MDL Number MFCD00052669
InChI Key DAEXXSXAEMFPHQ-UHFFFAOYNA-N
Synonym 2-4-chlorophenyl-3-oxopropanenitrile, 2-4-chlorophenyl-2-cyanoacetaldehyde, usaf el-67, alpha-formyl-p-chlorophenylacetonitrile, acetonitrile, 2-p-chlorophenyl-2-formyl, benzeneacetonitrile, 4-chloro-.alpha.-formyl, 3-10-00-03025 beilstein handbook reference, 4-chlorophenylcyanoacetaldehyde, benzeneacetonitrile,4-chloro-a-formyl, 2-formyl-2-4-chlorophenyl acetonitrile
PubChem CID 44090
IUPAC Name 2-(4-chlorophenyl)-3-oxopropanenitrile
SMILES ClC1=CC=C(C=C1)C(C=O)C#N

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Linear Formula ClC6H4CH(CN)CHO
Quantity 1 g
Formula Weight 179.61
Percent Purity 95%
Chemical Name or Material 2-(4-Chlorophenyl)-3-oxopropanenitrile
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.