accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COBrF

CAS RN 625446-22-2

IUPAC Name: 1-(4-bromo-2-fluorophenyl)ethan-1-one
Synonyms: 1-(4-bromo-2-fluorophenyl)ethanone
Molecular Weight (g/mol): 217.04
Molecular Formula: C8H6BrFO
InChi Key: ASKFCSCYGAFWAB-UHFFFAOYSA-N
SMILES: CC(=O)C1=C(F)C=C(Br)C=C1
Molecular Weight (g/mol) 
  • (5)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results