Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Bromo-2-fluoroacetophenone

Catalog No. SYX003071G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SYX003071G 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SYX003071G Supplier Sigma Aldrich Supplier No. SYX003071G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 3-chloro-2-butanone when treated with aq
  • Sodium thiocyanate forms 3-thiocyano-2-butanone.3-chloro-2-butanone was used to study various carbonylic compounds as substrates for 4-hydroxyacetophenone monooxygenase from pseudomonas fluorescens acb.prices for bulk quantities on request

Specifications

CAS 625446-22-2
Molecular Formula C8H6BrFO
Linear Formula C8H6BrFO
MDL Number MFCD03411551
Quantity 1 kg
Molecular Weight (g/mol) 217.04
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.