accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 626-22-2

IUPAC Name: 2-[3-(cyanomethyl)phenyl]acetonitrile
Synonyms: acetonitrile, m-phenylenedi-
Molecular Weight (g/mol): 156.19
Molecular Formula: C10H8N2
InChi Key: GRPFZJNUYXIVSL-UHFFFAOYSA-N
SMILES: N#CCC1=CC(CC#N)=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results