Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

m-Xylylene Dicyanide 98.0+%, TCI America™
SDP

Catalog No. X000225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
X000225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. X000225G Supplier TCI America Supplier No. X000225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 626-22-2
Molecular Formula C10H8N2
Molecular Weight (g/mol) 156.19
MDL Number MFCD00001915
InChI Key GRPFZJNUYXIVSL-UHFFFAOYSA-N
Synonym 1,3-phenylenediacetonitrile, 1,3-benzenediacetonitrile, m-phenylenediacetonitrile, m-benzenediacetonitrile, acetonitrile, m-phenylenedi, 2,2'-1,3-phenylene diacetonitrile, m-xylylene dicyanide, 1,3-bis cyanomethyl benzene, m-bis cyanomethyl benzene, 2-3-cyanomethyl phenyl acetonitrile
PubChem CID 69375
IUPAC Name 2-[3-(cyanomethyl)phenyl]acetonitrile
SMILES N#CCC1=CC(CC#N)=CC=C1

Specifications

Quantity 25 g
UN Number 3439
Formula Weight 156.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material m-Xylylene Dicyanide
RTECSNumber AM1780000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.