accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3NHCH3CH3

CAS RN 626-23-3

IUPAC Name: bis(butan-2-yl)amine
Synonyms: bis(sec-butyl)amine
Molecular Weight (g/mol): 129.25
Molecular Formula: C8H19N
InChi Key: OBYVIBDTOCAXSN-UHFFFAOYNA-N
SMILES: CCC(C)NC(C)CC
Boiling Point 
  • (2)
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)