Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Di-sec-butylamine, (+/-) + meso, 98%, Thermo Scientific™

Catalog No. AAB2569018
Change view
Click to view available options
Quantity:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2569018 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2569018 Supplier Thermo Scientific Chemicals Supplier No. B2569018

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 626-23-3
Boiling Point 134°C to 136°C
Flash Point 24°C (75°F)
Odor Amine-like
Molecular Formula C8H19N
Quantity 50g
MDL Number MFCD00009326
UN Number UN2733
Synonym di-sec-butylamine, mpba, 2-butanamine, n-1-methylpropyl, bis 1-methylpropyl amine, n-1-methylpropyl-2-butanamine, di-sec.-butylamine, bis methylpropyl amine, di-s-butylamine, di-sec.butylamine, bis butan-2-yl amine
InChI Key OBYVIBDTOCAXSN-UHFFFAOYSA-N
SMILES CCC(C)NC(C)CC
IUPAC Name N-butan-2-ylbutan-2-amine
Molecular Weight (g/mol) 129.247
PubChem CID 12277
Formula Weight 129.25
Percent Purity 98%
Chemical Name or Material Di-sec-butylamine, (+/-) + meso
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
H225-H314-H302-H312-H332
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.