accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 626-40-4

IUPAC Name: 3,5-dibromoaniline
Synonyms: 3,5-dibromoaniline
Molecular Weight (g/mol): 250.92
Molecular Formula: C6H5Br2N
InChi Key: RVNUUWJGSOHMRR-UHFFFAOYSA-N
SMILES: NC1=CC(Br)=CC(Br)=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)