Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dibromoaniline, 98%

Catalog No. AC460560250
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC460560250 25 g
AC460560050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC460560250 Supplier Thermo Scientific Chemicals Supplier No. 460560250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 626-40-4 | C6H5Br2N | 250.92 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 626-40-4
Molecular Formula C6H5Br2N
Molecular Weight (g/mol) 250.92
InChI Key RVNUUWJGSOHMRR-UHFFFAOYSA-N
Synonym 3,5-dibromophenylamine, 3,5-dibromanilin, benzenamine, 3,5-dibromo, 1-amino-3,5-dibromobenzene, 3,5-dibromobenzeneamine, 3,5-dibromoamiline, 3,5-di-bromo-aniline, 3,5-dibromo-phenylamine, acmc-1b0ij, ksc493e8l
PubChem CID 221512
IUPAC Name 3,5-dibromoaniline
SMILES C1=C(C=C(C=C1Br)Br)N

Specifications

Melting Point 53.0°C to 54.5°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Quantity 25 g
Formula Weight 250.92
Percent Purity 98%
Chemical Name or Material 3,5-Dibromoaniline
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. If skin irritation occurs: Get medical advice/attention. If eye irritation persists: Get medical advice/attention. Wash face, hands and any exposed skin thoroughly after handling.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.