accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHOH

CAS RN 626-95-9

IUPAC Name: pentane-1,4-diol
Synonyms: 1,4-dihydroxypentane
Molecular Weight (g/mol): 104.15
Molecular Formula: C5H12O2
InChi Key: GLOBUAZSRIOKLN-UHFFFAOYNA-N
SMILES: CC(O)CCCO
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results