accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 62613-15-4

IUPAC Name: λ⁵-arsenic(5+) diphenyliodanium hexafluoride
Synonyms: λ⁵-arsenic(5+) diphenyliodanium hexafluoride
Molecular Weight (g/mol): 470.03
Molecular Formula: C12H10AsF6I
InChi Key: ABPJIMXUCFBSRQ-UHFFFAOYSA-H
SMILES: [F-].[F-].[F-].[F-].[F-].[F-].[As+5].[I+](C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results