Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenyliodonium Hexafluoroarsenate 98.0+%, TCI America™
SDP

Catalog No. D22481G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D22481G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D22481G Supplier TCI America Supplier No. D22481G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 62613-15-4
Molecular Formula C12H10AsF6I
Molecular Weight (g/mol) 470.028
MDL Number MFCD00191354
InChI Key KFGZTBBPOZNSHA-UHFFFAOYSA-N
PubChem CID 2737135
IUPAC Name diphenyliodanium;hexafluoroarsenic(1-)
SMILES C1=CC=C(C=C1)[I+]C2=CC=CC=C2.F[As-](F)(F)(F)(F)F

Specifications

Melting Point 130°C
Color White
Quantity 1 g
UN Number 3465
Formula Weight 470.03
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Diphenyliodonium Hexafluoroarsenate
RTECSNumber NN6655000
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.