accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrONN(E/Z)

CAS RN 62625-24-5

IUPAC Name: 2-bromo-1-[4-(2-phenyldiazen-1-yl)phenyl]ethan-1-one
Synonyms: 2-bromo-1-[4-(2-phenyldiazen-1-yl)phenyl]ethanone
Molecular Weight (g/mol): 303.16
Molecular Formula: C14H11BrN2O
InChi Key: ZXTGCMLCYPYCID-UHFFFAOYSA-N
SMILES: BrCC(=O)C1=CC=C(C=C1)N=NC1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results