Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Phenylazophenacyl Bromide 98.0+%, TCI America™
SDP

Catalog No. B06341G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B06341G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B06341G Supplier TCI America Supplier No. B06341G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 62625-24-5
Molecular Formula C14H11BrN2O
Molecular Weight (g/mol) 303.159
MDL Number MFCD00017875
InChI Key ZXTGCMLCYPYCID-UHFFFAOYSA-N
Synonym 2-Bromo-4′C-phenylazoacetophenone
PubChem CID 112886
IUPAC Name 2-bromo-1-(4-phenyldiazenylphenyl)ethanone
SMILES C1=CC=C(C=C1)N=NC2=CC=C(C=C2)C(=O)CBr

Specifications

Melting Point 104°C
Quantity 1 g
UN Number 1759
Formula Weight 303.16
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Phenylazophenacyl Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.