accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 62637-91-6

IUPAC Name: potassium 2,6-dibromo-4-[(3,5-dibromo-4-oxocyclohexa-2,5-dien-1-ylidene)[2-(ethoxycarbonyl)phenyl]methyl]benzen-1-olate
Synonyms: potassium 2,6-dibromo-4-[(3,5-dibromo-4-oxocyclohexa-2,5-dien-1-ylidene)[2-(ethoxycarbonyl)phenyl]methyl]benzenolate
Molecular Weight (g/mol): 700.06
Molecular Formula: C22H13Br4KO4
InChi Key: WCIQBKUTYDIBJC-UHFFFAOYSA-M
SMILES: [K+].CCOC(=O)C1=CC=CC=C1C(C1=CC(Br)=C([O-])C(Br)=C1)=C1C=C(Br)C(=O)C(Br)=C1
Quantity 
  • (3)
  • (1)
  • (1)
  • (2)
Color 
  • (1)
  • (3)
Grade 
  • (3)
Percent Purity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (6)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)