Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrabromophenolphthalein ethyl ester, potassium salt, hydrate, 97%, Thermo Scientific™

Catalog No. AC420080010
Change view
Click to view available options
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC420080010 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC420080010 Supplier Thermo Scientific Chemicals Supplier No. 420080010

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 62637-91-6
Assay Percent Range 97%
Molecular Formula C22H13Br4KO4
MDL Number MFCD00011662
Merck Index 15, 9331
InChI Key WCIQBKUTYDIBJC-UHFFFAOYSA-M
SMILES [K+].CCOC(=O)C1=CC=CC=C1C(C1=CC(Br)=C([O-])C(Br)=C1)=C1C=C(Br)C(=O)C(Br)=C1
Infrared Spectrum Authentic
IUPAC Name potassium 2,6-dibromo-4-[(3,5-dibromo-4-oxocyclohexa-2,5-dien-1-ylidene)[2-(ethoxycarbonyl)phenyl]methyl]benzen-1-olate
Molecular Weight (g/mol) 700.06
PubChem CID 23689366
Formula Weight 700.05
Percent Purity ≥96.0%
Color Black-Blue to Blue
Quantity 1g
Chemical Name or Material Tetrabromophenolphthalein ethyl ester, potassium salt, hydrate, 97%
Show More Show Less
EINECSNumber 263-661-6
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.