accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6264-66-0

IUPAC Name: 4-(4-aminophenoxy)benzene-1,2-diamine
Synonyms: 4-(4-aminophenoxy)benzene-1,2-diamine
Molecular Weight (g/mol): 215.26
Molecular Formula: C12H13N3O
InChi Key: MPKIJEUTPZPJFP-UHFFFAOYSA-N
SMILES: NC1=CC=C(OC2=CC=C(N)C(N)=C2)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results