Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4,4'-Triaminodiphenyl Ether 98.0+%, TCI America™
SDP

Catalog No. T15615G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T15615G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T15615G Supplier TCI America Supplier No. T15615G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6264-66-0
Molecular Formula C12H13N3O
Molecular Weight (g/mol) 215.256
MDL Number MFCD00191504
InChI Key MPKIJEUTPZPJFP-UHFFFAOYSA-N
PubChem CID 80434
IUPAC Name 4-(4-aminophenoxy)benzene-1,2-diamine
SMILES C1=CC(=CC=C1N)OC2=CC(=C(C=C2)N)N

Specifications

Quantity 5 g
Formula Weight 215.26
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3,4,4′-Triaminodiphenyl Ether
RTECSNumber SS8080000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.