CAS RN 62658-64-4
IUPAC Name:
1-(tert-butylamino)-3-[(2-methyl-1H-indol-4-yl)oxy]propan-2-yl benzoate; propanedioic acid
Synonyms:
bopindolol; malonic acid
Molecular Weight (g/mol):
484.55
Molecular Formula:
C26H32N2O7
InChi Key:
LDOONRJGLKHGJI-UHFFFAOYNA-N
SMILES:
OC(=O)CC(O)=O.CC1=CC2=C(OCC(CNC(C)(C)C)OC(=O)C3=CC=CC=C3)C=CC=C2N1
1
–
1
of
1
results
1
Promotions Available
Thermo Scientific Chemicals Bopindolol malonate, 97+%
Thermo Scientific Chemicals Bopindolol malonate, 97+%
CAS: 62658-64-4 Molecular Formula: C23H28N2O3.C3H4O4