accessibility menu, dialog, popup

UserName

Viewing profile as user:
HO(E)

CAS RN 62668-02-4

IUPAC Name: (2E)-1,3-diphenylprop-2-en-1-ol
Synonyms: (2E)-1,3-diphenylprop-2-en-1-ol
Molecular Weight (g/mol): 210.28
Molecular Formula: C15H14O
InChi Key: ORACYDGVNJGDMI-VAWYXSNFNA-N
SMILES: OC(\C=C\C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results