CAS RN 62669-70-9
IUPAC Name:
3,6-diamino-9-[2-(methoxycarbonyl)phenyl]-10λ⁴-xanthen-10-ylium chloride
Synonyms:
rhodamine 123
Molecular Weight (g/mol):
380.83
Molecular Formula:
C21H17ClN2O3
InChi Key:
TUFFYSFVSYUHPA-UHFFFAOYSA-M
SMILES:
[Cl-].COC(=O)C1=CC=CC=C1C1=C2C=CC(N)=CC2=[O+]C2=CC(N)=CC=C12
1
–
1
of
1
results
1
Promotions Available
Thermo Scientific Chemicals Rhodamine 123, 99+%, pure
Thermo Scientific Chemicals Rhodamine 123, 99+%, pure
CAS: 62669-70-9 Molecular Formula: C21H19ClN2O4 Molecular Weight (g/mol): 398.843 MDL Number: MFCD00012664 InChI Key: SEXIURXGNASJRB-UHFFFAOYSA-N PubChem CID: 16211830 IUPAC Name: methyl 2-(3-amino-6-iminoxanthen-9-yl)benzoate;hydrate;hydrochloride SMILES: COC(=O)C1=CC=CC=C1C2=C3C=CC(=N)C=C3OC4=C2C=CC(=C4)N.O.Cl